ENAH93E74F19 Enamine

tert-butyl N-[(1S)-2-oxocyclopentyl]carbamate

  • MDL number MFCD18831387
  • Cas Number : 145106-46-3
  • Molecular Weight : 199.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7210056

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7210056
Availability details for
Quantity
  • Availability
  • Add to Cart