ENAH93E75024 Enamine

[3-(aminomethyl)azetidin-3-yl]methanamine trihydrochloride

  • Molecular Weight : 224.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name azetidine-3,3-diyldimethanamine trihydrochloride

Description

Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7563931
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart