ENAH93E7556D Enamine

(1R)-1-(2,5-difluorophenyl)ethan-1-amine hydrochloride

  • MDL number MFCD12757705
  • Cas Number : 1391449-47-0
  • Molecular Weight : 193.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1R)-1-(2,5-difluorophenyl)ethanamine hydrochloride
Inchi Code 1S/C8H9F2N.ClH/c1-5(11)7-4-6(9)2-3-8(7)10;/h2-5H,11H2,1H3;1H/t5-;/m1./s1
Inchi Key NQFUIZOIKVFLIM-NUBCRITNSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7832017

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7832017
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart