ENAH93E759D5 Enamine

N1-(1H-1,3-benzodiazol-2-yl)benzene-1,4-diamine

  • MDL number MFCD00569560
  • Cas Number : 133929-18-7
  • Molecular Weight : 224.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236-237
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-13113832

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13113832
Availability details for
Quantity
  • Availability
  • Add to Cart