ENAH93E75A1A Enamine

5-(bromomethyl)-2-methylpyrimidin-4-amine hydrobromide

  • MDL number MFCD05149311
  • Cas Number : 2908-71-6
  • Molecular Weight : 282.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-23609700

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23609700
Availability details for
Quantity
  • Availability
  • Add to Cart