ENAH942F28F5 Enamine

1-(4-chlorophenyl)-3-[(2R)-1-hydroxybutan-2-yl]urea

  • Molecular Weight : 242.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6569089

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6569089
Availability details for
Quantity
  • Availability
  • Add to Cart