ENAH942F2AA9 Enamine

1-{5-bromo-1H-pyrrolo[2,3-b]pyridin-3-yl}ethan-1-one

  • MDL number MFCD12403936
  • Cas Number : 866545-96-2
  • Molecular Weight : 239.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-268758

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-268758
Availability details for
Quantity
  • Availability
  • Add to Cart