ENAH942F2B2D Enamine

5-bromo-1,2-thiazole-3-carboxylic acid

  • MDL number MFCD24480225
  • Cas Number : 4576-89-0
  • Molecular Weight : 208.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179-182
Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6730887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730887
Availability details for
Quantity
  • Availability
  • Add to Cart