ENAH942F2B8C Enamine

3-(2,5-dimethyl-1H-pyrrol-1-yl)phenol

  • MDL number MFCD01314064
  • Cas Number : 97608-33-8
  • Molecular Weight : 187.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-72
Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7423636

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7423636
Availability details for
Quantity
  • Availability
  • Add to Cart