ENAH942F2E0C Enamine

4-(trifluoromethyl)-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD13190112
  • Cas Number : 149427-58-7
  • Molecular Weight : 179.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-105341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105341
Availability details for
Quantity
  • Availability
  • Add to Cart