ENAH942F2E16 Enamine

(2S)-3-(3-fluorophenyl)-2-hydroxypropanoic acid

  • MDL number MFCD28977372
  • Cas Number : 159415-52-8
  • Molecular Weight : 184.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7411719

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7411719
Availability details for
Quantity
  • Availability
  • Add to Cart