ENAH942F2EB5 Enamine

1-chloro-2-iodo-5-methoxy-4-nitrobenzene

  • MDL number MFCD27978470
  • Cas Number : 1508278-48-5
  • Molecular Weight : 313.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-8309527

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-8309527
Availability details for
Quantity
  • Availability
  • Add to Cart