ENAH942F2F75 Enamine

methyl 6-methoxy-2-methylpyridine-3-carboxylate

  • MDL number MFCD16611640
  • Cas Number : 137347-39-8
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7400056

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7400056
Availability details for
Quantity
  • Availability
  • Add to Cart