ENAH9453D409 Enamine

(3R,5R)-5-(aminomethyl)pyrrolidin-3-ol

  • MDL number MFCD32199727
  • Cas Number : 2165898-75-7
  • Molecular Weight : 116.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6935380

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6935380
Availability details for
Quantity
  • Availability
  • Add to Cart