ENAH9453D510 Enamine

1-phenylazetidin-3-ol

  • MDL number MFCD22566181
  • Cas Number : 857280-53-6
  • Molecular Weight : 149.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-196411

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196411
Availability details for
Quantity
  • Availability
  • Add to Cart