ENAH9453D54F Enamine

8-fluoroquinoline-4-carbaldehyde

  • MDL number MFCD06824407
  • Cas Number : 496792-30-4
  • Molecular Weight : 175.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7399624

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7399624
Availability details for
Quantity
  • Availability
  • Add to Cart