ENAH9453D5A1 Enamine

5-fluoro-2,3-dihydro-1-benzofuran-7-carbaldehyde

  • MDL number MFCD07368090
  • Cas Number : 245762-36-1
  • Molecular Weight : 166.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-105194

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105194
Availability details for
Quantity
  • Availability
  • Add to Cart