ENAH9453D6E7 Enamine

2-(1-aminoethyl)-4-methyl-1,3-oxazole-5-carboxylic acid

  • MDL number MFCD24513865
  • Cas Number : 1822606-28-9
  • Molecular Weight : 170.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7894076

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7894076
Availability details for
Quantity
  • Availability
  • Add to Cart