ENAH9453D8D9 Enamine

3-iodo-2,6-dimethoxypyridine

  • MDL number MFCD09608101
  • Cas Number : 214360-56-2
  • Molecular Weight : 265.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7721371

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7721371
Availability details for
Quantity
  • Availability
  • Add to Cart