ENAH9453D929 Enamine

2,6-difluoro-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

  • MDL number MFCD29088262
  • Molecular Weight : 268.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-21665214

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-21665214
Availability details for
Quantity
  • Availability
  • Add to Cart