ENAH9453DA2E Enamine

tert-butyl N-(6-amino-2,3-dihydro-1H-inden-1-yl)carbamate

  • MDL number MFCD32200709
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6856158

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6856158
Availability details for
Quantity
  • Availability
  • Add to Cart