ENAH9453DAC4 Enamine

2-(3-bromo-2-chlorophenyl)acetic acid

  • MDL number MFCD18390581
  • Cas Number : 1261438-67-8
  • Molecular Weight : 249.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1720146

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720146
Availability details for
Quantity
  • Availability
  • Add to Cart