ENAH9453DAD1 Enamine

2-{[(tert-butoxy)carbonyl](2-phenylethyl)amino}acetic acid

  • MDL number MFCD24468123
  • Molecular Weight : 279.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1071922

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1071922
Availability details for
Quantity
  • Availability
  • Add to Cart