ENAH9453DAE5 Enamine

2-[2-methoxy-5-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD22414483
  • Cas Number : 1688686-12-5
  • Molecular Weight : 302.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-23704489

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23704489
Availability details for
Quantity
  • Availability
  • Add to Cart