ENAH9453DAF6 Enamine

2-(3-ethoxy-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD25102229
  • Cas Number : 1416367-03-7
  • Molecular Weight : 266.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-4308689

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-4308689
Availability details for
Quantity
  • Availability
  • Add to Cart