ENAH9453DB1F Enamine

2-(4-chloro-3,5-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD22418269
  • Cas Number : 1165935-96-5
  • Molecular Weight : 274.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-12578623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12578623
Availability details for
Quantity
  • Availability
  • Add to Cart