ENAH9453DB75 Enamine

5-chloro-3-methyl-3H-imidazo[4,5-b]pyridine

  • MDL number MFCD11047120
  • Cas Number : 83472-65-5
  • Molecular Weight : 167.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6473544

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6473544
Availability details for
Quantity
  • Availability
  • Add to Cart