ENAH9453DBA5 Enamine

1-methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylic acid

  • MDL number MFCD11043031
  • Cas Number : 934568-20-4
  • Molecular Weight : 176.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-3048432

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-3048432
Availability details for
Quantity
  • Availability
  • Add to Cart