ENAH9453DCF7 Enamine

sodium 5-(4-methoxyphenyl)-1,3,4-oxadiazole-2-carboxylate

  • Molecular Weight : 242.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-27103694

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27103694
Availability details for
Quantity
  • Availability
  • Add to Cart