ENAH9453DE4A Enamine

4-(oxetan-3-yl)aniline

  • MDL number MFCD20040419
  • Cas Number : 1221819-62-0
  • Molecular Weight : 149.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-378587

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-378587
Availability details for
Quantity
  • Availability
  • Add to Cart