ENAH9453DE4E Enamine

(2R,3S,4R,5R)-2-(hydroxymethyl)-5-(9H-purin-9-yl)oxolane-3,4-diol

  • MDL number MFCD00022819
  • Cas Number : 550-33-4
  • Molecular Weight : 252.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-184
Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7400810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7400810
Availability details for
Quantity
  • Availability
  • Add to Cart