ENAH9453DE5E Enamine

2-methyl-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid

  • MDL number MFCD18253774
  • Cas Number : 933717-06-7
  • Molecular Weight : 176.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-4301100

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-4301100
Availability details for
Quantity
  • Availability
  • Add to Cart