ENAH9453DF1C Enamine

dibenzyl(nitroso)amine

  • MDL number MFCD00019921
  • Cas Number : 5336-53-8
  • Molecular Weight : 226.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7281779

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7281779
Availability details for
Quantity
  • Availability
  • Add to Cart