ENAH9453E2A7 Enamine

4-(azetidin-3-yl)pyrimidine dihydrochloride

  • MDL number MFCD22573701
  • Cas Number : 1384264-47-4
  • Molecular Weight : 208.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7422003

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7422003
Availability details for
Quantity
  • Availability
  • Add to Cart