ENAH9453E2F6 Enamine

3-(3-methoxyphenyl)pyrrolidine hydrochloride

  • MDL number MFCD03701257
  • Cas Number : 1095545-66-6
  • Molecular Weight : 213.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7416733

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7416733
Availability details for
Quantity
  • Availability
  • Add to Cart