FRO026586948 Frontier Scientific Services Inc.

2-[(2-Chlorophenyl)Methyl]Pyrrolidine

  • MDL number MFCD02663525
  • Cas Number : 91564-44-2
  • Molecular Weight : 195.688995

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart