FRO026587753 Frontier Scientific Services Inc.

3,5-Dinitrophenyl Isocyanate

  • MDL number MFCD00210046
  • Cas Number : 59776-60-2
  • Molecular Weight : 209.1191

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart