FRO026587852 Frontier Scientific Services Inc.

4-N-Butoxyphenyl Isocyanate

  • MDL number MFCD00037042
  • Cas Number : 28439-86-3
  • Molecular Weight : 191.2318

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart