FRO026588207 Frontier Scientific Services Inc.

3,4-DIMETHYL-6-ETHYLPHENOL

  • MDL number MFCD00020107
  • Cas Number : 2219-78-5
  • Molecular Weight : 150.2225

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data FREE
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart