FRO026588263 Frontier Scientific Services Inc.

Phloroglucinol Aldehyde Triethyl Ether

  • MDL number MFCD00016586
  • Cas Number : 59652-88-9
  • Molecular Weight : 238.28601

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart