FRO026588264 Frontier Scientific Services Inc.

2-Chloro-4-Phenylphenol

  • MDL number MFCD00045745
  • Cas Number : 92-04-6
  • Molecular Weight : 204.651993

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart