FRO026588651 Frontier Scientific Services Inc.

3-(PENTYLAMINO)PROPIONITRILE

  • MDL number MFCD00070654
  • Cas Number : 59676-91-4
  • Molecular Weight : 140.2301

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data FREE
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart