FRO026588787 Frontier Scientific Services Inc.

5-(CHLOROMETHYL)MECONIN

  • MDL number MFCD00023102
  • Cas Number : 29809-17-4
  • Molecular Weight : 242.6609

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data FREE
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart