FRO026589114 Frontier Scientific Services Inc.

5-(3,4-Dichlorophenyl)-1H-Tetrazole

  • MDL number MFCD01075710
  • Cas Number : 41421-27-6
  • Molecular Weight : 215.0426

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart