FRO026589142 Frontier Scientific Services Inc.

3,4-Dimethylphenylacetonitrile

  • MDL number MFCD00019829
  • Cas Number : 3020-06-2
  • Molecular Weight : 145.2059

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart