FRO026589764 Frontier Scientific Services Inc.

2-Ethoxynaphthalene-1-Carbonyl Chloride

  • MDL number MFCD00046426
  • Cas Number : 55150-29-3
  • Molecular Weight : 234.677994

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart