FRO026590127 Frontier Scientific Services Inc.

2-CYCLOHEXYLAMINO-1-PHENYLETHANOL

  • MDL number MFCD00021321
  • Cas Number : 6589-48-6
  • Molecular Weight : 219.32957

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data FREE
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart