FRO026590411 Frontier Scientific Services Inc.

N-(4-Amino-5-Methoxy-2-Methylphenyl)Benzamide

  • MDL number MFCD00064435
  • Cas Number : 99-21-8
  • Molecular Weight : 256.299713

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart