FRO026590556 Frontier Scientific Services Inc.

3,4'-Dichloropropiophenone

  • MDL number MFCD00000992
  • Cas Number : 3946-29-0
  • Molecular Weight : 203.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart