FRO026591335 Frontier Scientific Services Inc.

N-CarbamoYl-Maleamic Acid

  • MDL number MFCD00014790
  • Cas Number : 105-61-3
  • Molecular Weight : 158.1146

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart